Triphenylphosphine oxide ≥98%

T84603-100G T84603-25G
220228-085EA 48.94 USD
220228-085 220228-086
Triphenylphosphine oxide ≥98%
Triphenylphosphine oxide
Formula: C₁₈H₁₅OP
MW: 278.29 g/mol
Boiling Pt: ? 360 °C (1013 hPa)
Melting Pt: 155 - 158 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: 180 °C
MDL Number: MFCD00002080
CAS Number: 791-28-6
EINECS: 212-338-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR