DL-Threonine ≥98.5%

T8375-1KG T8375-100G
220228-050EA 389.69 USD
220228-050 220228-049
DL-Threonine ≥98.5%
DL-Threonine
Formula: C₄H₉NO₃
MW: 119.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063722
CAS Number: 80-68-2
EINECS: 201-300-6

Order Now

Specification Test Results

Assay 98.5% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR