Trityl bromide ≥98%

Synonyms: Bromotriphenylmethane, Triphenylbromomethane, Triphenylmethyl bromide

T83607-100G T83607-25G
220228-047EA 505.08 USD
220228-047 220228-048
Trityl bromide ≥98%
Trityl bromide
Formula: (C₆H₅)₃CBr
MW: 323.23 g/mol
Boiling Pt: 230 °C (1013 hPa)
Density: 1.55 g/cm³ (20 °C)
MDL Number: MFCD00000120
CAS Number: 596-43-0
EINECS: 209-884-4
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR