Thallium(I) acetate ≥99%

Danger

Synonyms: Acetic acid thallium (I) salt, Thallous acetate

T8266-100G T8266-25G T8266-5G
220228-022EA 860 USD
220228-022 220228-023 220228-024
Thallium(I) acetate ≥99%
Thallium(I) acetate
Formula: C₂H₃O₂Tl
MW: 263.43 g/mol
Boiling Pt: 1457 °C (1013 hPa)
Melting Pt: 110 °C
Density: 3.68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013045
CAS Number: 563-68-8
EINECS: 209-257-5
UN: 1707
ADR: 6.1,II
Merck Index: 12,09392

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR