D-Threonine ≥98%

Synonyms: H-D-Thr-OH, (2R,3S)-2-Amino-3-hydroxybutanoic acid

T8250-5G T8250-100G T8250-25G
220228-016EA 67.69 USD
220228-016 220228-014 220228-015
D-Threonine ≥98%
D-Threonine
Formula: C₄H₉NO₃
MW: 119.12 g/mol
MDL Number: MFCD00064269
CAS Number: 632-20-2
EINECS: 211-171-8

Order Now

Specification Test Results

Assay 98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR