Triphenylamine ≥98%

T81604-100G T81604-25G
220225-526EA 100.8 USD
220225-526 220225-527
Triphenylamine ≥98%
Triphenylamine
Formula: C₁₈H₁₅N
MW: 245.32 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 123 - 128 °C
Density: 0.774 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003020
CAS Number: 603-34-9
EINECS: 210-035-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR