Triphenylacetic acid ≥99%

Synonyms: 2,2,2-Triphenylacetic acid

T81205-5G T81205-10G
220226-601EA 79.58 USD
220226-601 220226-600
Triphenylacetic acid ≥99%
Triphenylacetic acid
Formula: C₂₀H₁₆O₂
MW: 288.35 g/mol
Boiling Pt: 437 °C (1013 hPa)
Melting Pt: 267 - 273 °C
Density: 1.27 g/cm³ (20 °C)
MDL Number: MFCD00004185
CAS Number: 595-91-5
EINECS: 209-873-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR