2-Thiouracil

Synonyms: 4-Hydroxypyrimidine-2(1H)-thione

T7750-250G T7750-100G T7750-1KG T7750-25G
220227-945EA 306.92 USD
220227-945 220227-943 220227-944 220227-946
2-Thiouracil
2-Thiouracil
Formula: C₄H₄N₂OS
MW: 128.15 g/mol
Melting Pt: 340 °C
MDL Number: MFCD00006039
CAS Number: 141-90-2
EINECS: 205-508-8
Merck Index: 12,09504

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR