3,3,5-Trimethylcyclohexanone ≥98%

T75752-250ML T75752-1L
220227-907EA 40.28 USD
220227-907 220227-906
3,3,5-Trimethylcyclohexanone ≥98%
3,3,5-Trimethylcyclohexanone
Formula: C₉H₁₆O
MW: 140.23 g/mol
Boiling Pt: 188.8 °C (1013 hPa)
Melting Pt: –10 °C
Density: 0.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00019466
CAS Number: 873-94-9
EINECS: 212-855-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR