Tetramethylammonium hydroxide pentahydrate

Danger

T7505-5G T7505-1KG T7505-25G T7505-100G
220227-881EA 56.35 USD
220227-881 220227-879 220227-880 220227-878
Tetramethylammonium hydroxide pentahydrate
Tetramethylammonium hydroxide pentahydrate
Formula: C₄H₁₃NO·5H₂O
Melting Pt: 65…68 °C
Density: 1.829 g/cm³ (20 °C)
MDL Number: MFCD00149566
CAS Number: 10424-65-4
EINECS: 200-882-9
UN: 3423
ADR: 8,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR