N,N,N',N'-Tetramethyl-p-phenylenediamine

Warning

Synonyms: N,N,N,N-Tetramethyl-1,4-phenylenediamine, N,N,N′,N′-Tetramethyl-p-phenylenediamine, TMPD, N,N,N',N'-Tetramethyl-1,4-phenylenediamine

T7394-25G T7394-5G
220227-856EA 295.08 USD
220227-856 220227-857
N,N,N',N'-Tetramethyl-p-phenylenediamine
N,N,N',N'-Tetramethyl-p-phenylenediamine
Formula: C₁₀H₁₆N₂
MW: 164.25 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 48…50 °C
Density: 1.08 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008309
CAS Number: 100-22-1
EINECS: 202-831-6
UN: 1673
ADR: 6.1,III
Merck Index: 12,09367

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR