Pivaloyl chloride ≥99%

Synonyms: Trimethylacetyl chloride

T72605-100ML T72605-500ML T72605-5ML
220227-824EA 33.49 USD
220227-824 220227-825 220227-826
Pivaloyl chloride ≥99%
Pivaloyl chloride
Formula: C₅H₉ClO
MW: 120.58 g/mol
Boiling Pt: 105 °C (1013 hPa)
Melting Pt: -56 °C
Density: 0.979 g/cm³ (20 °C)
Flash Pt: 8 °C
Storage Temperature: Ambient
MDL Number: MFCD00000709
CAS Number: 3282-30-2
EINECS: 221-921-6
UN: 2438
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR