Pivalaldehyde (trimethylacetaldehyde) ≥96%

Synonyms: Pivalaldehyde, Pivaldehyde, Trimethylacetaldehyde, 2,2-Dimethylpropionaldehyde, 2,2-Dimethylpropanal

T71501-100ML T71501-25ML T71501-5ML
220226-582EA 624.62 USD
220226-582 220226-583 220226-584
Pivalaldehyde (trimethylacetaldehyde) ≥96%
Pivalaldehyde (trimethylacetaldehyde)
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 74 °C (1013 hPa)
Melting Pt: 6 °C
Density: 0.771 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00006962
CAS Number: 630-19-3
EINECS: 211-134-6

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR