(3,4,5-Trimethoxyphenyl)acetic acid ≥97%

Synonyms: 3,4,5-Trimethoxyphenylacetic acid

T70602-50G T70602-10G
220227-788EA 287.54 USD
220227-788 220227-787
(3,4,5-Trimethoxyphenyl)acetic acid ≥97%
(3,4,5-Trimethoxyphenyl)acetic acid
Formula: (C₆H₂(OCH₃)₃)CH₂COOH
MW: 226.23 g/mol
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 117…120 °C
MDL Number: MFCD00004336
CAS Number: 951-82-6
EINECS: 213-456-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR