3,4,5-Trimethoxybenzoic acid ≥99%

Synonyms: Trimethylgallic Acid

T69000-100G T69000-5G
220227-195EA 76.03 USD
220227-195 220227-196
3,4,5-Trimethoxybenzoic acid ≥99%
3,4,5-Trimethoxybenzoic acid
Formula: C₁₀H₁₂O₅
MW: 212.2 g/mol
Boiling Pt: 376 °C (1013 hPa)
Melting Pt: 168…170 °C
Density: 1.31 g/cm³ (24.95 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002501
CAS Number: 118-41-2
EINECS: 204-248-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR