Triisopropyl phosphite ≥95%

Synonyms: Tri-i-propylphosphite

T67806-500ML T67806-100ML
220227-732EA 196 USD
220227-732 220227-731
Triisopropyl phosphite ≥95%
Triisopropyl phosphite
Formula: C₉H₂₁O₃P
MW: 208.24 g/mol
Boiling Pt: 63 °C (1013 hPa)
Density: 0.9361 g/cm³ (0 °C)
MDL Number: MFCD00008874
CAS Number: 116-17-6
EINECS: 204-130-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR