1,1,1-Trifluoroacetone ≥97%

T62804-5G T62804-25G T62804-100G
220227-526EA 47.17 USD
220227-526 220227-525 220227-524
1,1,1-Trifluoroacetone ≥97%
1,1,1-Trifluoroacetone
Formula: C₃H₃F₃O
MW: 112.05 g/mol
Boiling Pt: 22 °C (1013 hPa)
Melting Pt: -78 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000423
CAS Number: 421-50-1
EINECS: 207-005-9
UN: 1993
ADR: 3,I

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR