Triethyl phosphite ≥98%

T61204-4L T61204-5ML T61204-100ML T61204-500ML
220227-452EA 141.92 USD
220227-452 220227-499 220227-451 220227-453
Triethyl phosphite ≥98%
Triethyl phosphite
Formula: C₆H₁₅O₃P
MW: 166.16 g/mol
Boiling Pt: 156 °C (1013 hPa)
Melting Pt: 51 °C
Density: 0.9643 g/cm³ (20 °C)
Flash Pt: 52 °C
MDL Number: MFCD00009084
CAS Number: 122-52-1
EINECS: 204-552-5
UN: 2323
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR