Triethanolamine (trolamine) ≥98%, Reagent Grade

Synonyms: Tris-(2-hydroxyethyl)amine, 2,2',2''-Nitrilotriethanol

T58300-20KG T58300-4KG T58300-1KG T58300-25G
220227-379EA 830.77 USD
220227-379 220227-381 220227-378 220227-380
Triethanolamine (trolamine) ≥98%, Reagent Grade
Triethanolamine (trolamine)
Formula: C₆H₁₅NO₃
MW: 149.19 g/mol
Boiling Pt: 335.4 °C (1013 hPa)
Melting Pt: 21.6 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 179 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002855
CAS Number: 102-71-6
EINECS: 203-049-8
Merck Index: 13,09739
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c181d3e80375aec7c34e17766247484e1c3d3257&vid=40c3d3d0c0162ac22208cadb8d5254a7cf0b340a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR