2,4,6-Trichlorophenol ≥98%

Warning

T55301-250G T55301-50G T55301-1KG
220227-303EA 109.85 USD
220227-303 220227-304 220227-302
2,4,6-Trichlorophenol ≥98%
2,4,6-Trichlorophenol
Formula: C₆H₃Cl₃O
MW: 197.45 g/mol
Boiling Pt: 244.5 °C (1013 hPa)
Melting Pt: ∼66 °C
Density: 1.675 g/cm³ (25 °C)
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00002172
CAS Number: 88-06-2
EINECS: 201-795-9
UN: 2020
ADR: 6.1,III
Merck Index: 12,09773

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR