2-Thiobarbituric acid

Synonyms: 4,6-Dihydroxy-2-mercaptopyrimidine

T5500-25G T5500-500G T5500-100G T5500-250G
220227-288EA 57 USD
220227-288 220227-289 220227-286 220227-287
2-Thiobarbituric acid
2-Thiobarbituric acid
Formula: C₄H₄N₂O₂S
MW: 144.15 g/mol
Boiling Pt: 419 °C (1013 hPa)
Melting Pt: 235 °C
Density: 1.661 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006674
CAS Number: 504-17-6
EINECS: 207-985-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR