2,2,2-Trichloroethanol ≥99%

Danger

Synonyms: 2,2,2-Trichloroethan-1-ol

T54801-100G T54801-500G
220227-284EA 47.69 USD
220227-284 220227-285
2,2,2-Trichloroethanol ≥99%
2,2,2-Trichloroethanol
Formula: C₂H₃Cl₃O
MW: 149.4 g/mol
Boiling Pt: 151 °C (1013 hPa)
Melting Pt: 17 - 18 °C
Density: 1.56 g/cm³ (20 °C)
Flash Pt: ≥ 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004677
CAS Number: 115-20-8
EINECS: 204-071-0
UN: 2810
ADR: 6.1,III
Merck Index: 12,09786

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR