Tributyltin chloride

Danger

Synonyms: Chlorotributylstannane, Tri-n-butyltin chloride, TBTC

T50202-500G T50202-5G T50202-100G
220227-071EA 138.46 USD
220227-071 220227-072 220227-070
Tributyltin chloride
Tributyltin chloride
Formula: C₁₂H₂₇ClSn
MW: 325.51 g/mol
Boiling Pt: 171…173 °C (25 mmHg)
Melting Pt: –9 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: >100 °C
MDL Number: MFCD00000521
CAS Number: 1461-22-9
EINECS: 215-958-7
UN: 2788
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR