Tetradecyltrimethylammonium bromide, Sigma-Aldrich®

Danger

Synonyms: MiTMAB, (1-Tetradecyl)trimethylammonium bromide, N,N,N-Trimethyltetradecan-1-aminium bromide, Tetradonium bromide, Tetradecyl trimethyl ammonium Bromide, Myristyltrimethylammonium bromide, Dynamin Inhibitor II, TTAB

T4762-500G T4762-250G T4762-100G T4762-1KG T4762-5G
220227-010EA 368.77 USD
220227-010 220225-770 220225-768 220225-769 220227-011
Tetradecyltrimethylammonium bromide, Sigma-Aldrich®
Tetradecyltrimethylammonium bromide
Formula: C₁₇H₃₈BrN
MW: 336.394 g/mol
Melting Pt: 245…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00011770
CAS Number: 1119-97-7
EINECS: 214-291-9
UN: 3261
ADR: 8,II
Merck Index: 12,06419

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR