Tetrazolium blue chloride

Synonyms: Blue Tetrazolium, 3,3′-(3,3′-Dimethoxy[1,1′-biphenyl]-4,4′-diyl)-bis(2,5-diphenyl-2H-tetrazolium) dichloride, 3,3'-(3,3'-Dimethoxy-4,4'-biphenylene)bis(2,5-diphenyl-2H-tetrazolium chloride), BTC, 3,3′-Dianisole-4,4′-bis(3,5-diphenyltetrazolium chloride), Blue Tetrazolium chloride

T4375-1G T4375-25G T4375-5G T4375-10G
220226-267EA 75.74 USD
220226-267 220226-268 220226-269 220226-266
Tetrazolium blue chloride
Tetrazolium blue chloride
Formula: C₄₀H₃₂Cl₂N₈O₂
MW: 727.65 g/mol
Melting Pt: 265 - 270 °C
Storage Temperature: Ambient
MDL Number: MFCD00040933
CAS Number: 1871-22-3
EINECS: 217-488-8
Merck Index: 12,09380
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=825ba12666da399ee548628826b021d3cbd4ba71&vid=825ba12666da399ee548628826b021d3cbd4ba71&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf42164c85c3ff11937c1db791fff066d2010ddb&vid=2fc65401f1ae23533e1bdd2b18b303813894aa03&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR