o-Tolyl isocyanate ≥99%

Synonyms: 2-Methylphenyl isocyanate, 1-Isocyanato-2-methylbenzene

T40703-10G T40703-50G
220226-898EA 109.85 USD
220226-898 220226-899
o-Tolyl isocyanate ≥99%
o-Tolyl isocyanate
Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 185 °C (1013 hPa)
Density: 1.07 g/cm³ (20 °C)
MDL Number: MFCD00002009
CAS Number: 614-68-6
EINECS: 210-389-0
UN: 3334
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR