Tetranitro blue tetrazolium

Synonyms: TNBT, 3,3'-(3,3'-Dimethoxy[1,1'-biphenyl]-4,4'-diyl)bis[2,5-bis(p-nitrophenyl)-2H-tetrazolium] dichloride

T4000-100MG T4000-500MG T4000-1G T4000-250MG T4000-50MG
220226-859EA 155.38 USD
220226-859 220226-862 220226-860 220226-861 220226-863
Tetranitro blue tetrazolium
Tetranitro blue tetrazolium
Formula: C₄₀H₂₈Cl₂N₁₂O₁₀
MW: 907.64 g/mol
Melting Pt: 165 °C
MDL Number: MFCD00036338
CAS Number: 1184-43-6
EINECS: 214-665-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9a651580deeb18ef2d0662f2c0bcf7e3530feef&vid=5557b1ca500fd385826c04951e761c90d7caa117&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR