2,4-Toluenediisocyanate ≥95%

Synonyms: 4-Methyl-m-phenylene diisocyanate, Tolylene 2,4-diisocyanate, 2,4-TDI

T39853-250G T39853-100G
220226-855EA 139.92 USD
220226-855 220226-854
2,4-Toluenediisocyanate ≥95%
2,4-Toluenediisocyanate
Formula: C₉H₆N₂O₂
MW: 174.16 g/mol
MDL Number: MFCD00002011
CAS Number: 584-84-9
EINECS: 209-544-5
UN: 2078
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR