p-Tolylacetic acid ≥99%

Synonyms: 2-(p-Tolyl)acetic acid, 4-Tolylacetic acid, (4-Methylphenyl)acetic acid

T38105-5G T38105-25G
220226-809EA 97.14 USD
220226-809 220226-808
p-Tolylacetic acid ≥99%
p-Tolylacetic acid
Formula: (C₆H₄(CH₃))CH₂COOH
MW: 150.18 g/mol
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 91 °C
MDL Number: MFCD00004353
CAS Number: 622-47-9
EINECS: 210-738-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR