o-Tolylacetic acid ≥99%

Synonyms: 2-Tolylacetic acid, (2-Methylphenyl)acetic acid

T38083-5G T38083-25G
220226-803EA 55.15 USD
220226-803 220226-802
o-Tolylacetic acid ≥99%
o-Tolylacetic acid
Formula: (C₆H₄CH₃)CH₂COOH
MW: 150.18 g/mol
Boiling Pt: 283 °C (1013 hPa)
Melting Pt: 86 - 87 °C
MDL Number: MFCD00004328
CAS Number: 644-36-0
EINECS: 211-416-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR