DL-α-Tocopherol acetate

T3376-100G T3376-25G T3376-5G
220226-684EA 169.4 USD
220226-684 220226-685 220226-686
DL-α-Tocopherol acetate
DL-α-Tocopherol acetate
Formula: C₃₁H₅₂O₃
MDL Number: MFCD00072042
CAS Number: 7695-91-2
EINECS: 231-710-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR