Thiosemicarbazide ≥99%

Danger

Synonyms: 3-Thiosemicarbazide

T33405-500G T33405-100G
220226-683EA 122.77 USD
220226-683 220226-682
Thiosemicarbazide ≥99%
Thiosemicarbazide
Formula: CH₅N₃S
MW: 91.14 g/mol
Melting Pt: 178…180 °C
MDL Number: MFCD00007620
CAS Number: 79-19-6
EINECS: 201-184-7
UN: 2811
ADR: 6.1,II
Merck Index: 12,09499

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR