Thiosalicylic acid ≥97%

Warning

Synonyms: 2-Thiobenzoic acid, 2-Mercaptobenzoic acid

T33200-100G T33200-5G T33200-500G
220226-672EA 65.26 USD
220226-672 220226-674 220226-673
Thiosalicylic acid ≥97%
Thiosalicylic acid
Formula: C₇H₆O₂S
MW: 154.19 g/mol
Boiling Pt: 230 °C (0.02 -rr)
Melting Pt: 165 - 166 °C
Density: 1.489 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004836
CAS Number: 147-93-3
EINECS: 205-704-3
UN: 3335
ADR: 9,
Merck Index: 12,09498

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR