Toluidine blue O

Synonyms: TBO, 3-Amino-7-(dimethylamino)-2-methyl-5-phenothiazinium chloride, 3-Amino-7-(dimethylamino)-2-methylphenothiazin-5-ium chloride, Tolonium chloride, Basic Blue 17, Blutene chloride

T3260-5G T3260-100G T3260-25G
220226-642EA 108.78 USD
220226-642 220226-640 220226-641
Toluidine blue O
Toluidine blue O
Formula: C₁₅H₁₆ClN₃S
MW: 305.83 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011934
CAS Number: 92-31-9
EINECS: 202-146-2
Merck Index: 12,09658
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b68e54e7aa7ac291c3a6c1e795308f18da86b2c&vid=60380f47bd5001b6d4c2dcdf8e15df28ba66eedf&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR