p-Terphenyl ≥99%

Synonyms: 4-Phenyl-1,1'-biphenyl, 1,4-Diphenylbenzene, 4-Phenylbiphenyl, 1,4-Terphenyl

T3203-100G T3203-25G
220226-561EA 215.77 USD
220226-561 220226-562
p-Terphenyl ≥99%
p-Terphenyl
Formula: C₁₈H₁₄
MW: 230.31 g/mol
Boiling Pt: 383 °C (1013 hPa)
Melting Pt: 210…211 °C
Density: 1.234 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003061
CAS Number: 92-94-4
EINECS: 202-205-2
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR