N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride

Warning

Synonyms: N1,N1,N4,N4-Tetramethylbenzene-1,4-diamine dihydrochloride, TMPD 2 HCl, N,N,N',N'-Tetramethyl-1,4-phenylenediamine dihydrochloride, N,N,N',N'-Tetramethyl-1,4-phenylenediammonium dichloride

T3134-25G T3134-5G
220226-539EA 340 USD
220226-539 220226-540
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride
Formula: C₁₀H₁₈Cl₂N₂
MW: 237.17 g/mol
Melting Pt: 227 - 230 °C
Storage Temperature: Ambient
MDL Number: MFCD00012482
CAS Number: 637-01-4
EINECS: 211-274-8
Merck Index: 12,09367
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cdb59462690da7f44f0ba4e2ee465bb1fa318e61&vid=f952549e50e1861aa87fb1b41ad0a9689a7cd94e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR