3,3'-Thiodipropionic acid ≥97%

Synonyms: 3,3-Thiodipropionic acid

T30201-100G T30201-3KG
220226-501EA 85.08 USD
220226-501 220226-502
3,3'-Thiodipropionic acid ≥97%
3,3'-Thiodipropionic acid
Formula: C₆H₁₀O₄S
MW: 178.21 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 128…131 °C
Density: 1.496 g/cm³ (20 °C)
MDL Number: MFCD00002781
CAS Number: 111-17-1
EINECS: 203-841-3
Merck Index: 12,09468

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR