Tetraphenylarsonium chloride monohydrate

Danger

Synonyms: Tetraphenylarsonium(V) chloride hydrate

T25305-10G T25305-2.5G
220225-822EA 431.31 USD
220225-822 220225-823
Tetraphenylarsonium chloride monohydrate
Tetraphenylarsonium chloride monohydrate
Formula: C₂₄H₂₀AsCl·1H₂O
MW: 436.81 g/mol
Melting Pt: 258 - 260 °C
MDL Number: MFCD00011908
CAS Number: 104170-16-3
EINECS: 208-070-6
UN: 3465
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR