Tetranitromethane

T25003-1G T25003-5G
220226-381EA 66.95 USD
220226-381 220226-382
Tetranitromethane
Tetranitromethane
Formula: CN₄O₈
MW: 196.03 g/mol
Boiling Pt: 126 °C (1013 hPa)
Melting Pt: 14.2 °C
Density: 1.629 g/cm³ (20 °C)
Storage Temperature: Refrigerator
CAS Number: 509-14-8
EINECS: 208-094-7
UN: 1510
ADR: 5.1,I
Merck Index: 12,09371

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR