Thiram ≥97%

Warning

Synonyms: Tetramethylthiuram disulphide

T24201-100G T24201-5G
220226-363EA 28.86 USD
220226-363 220226-364
Thiram ≥97%
Thiram
Formula: C₆H₁₂N₂S₄
MW: 240.44 g/mol
Boiling Pt: 307 °C (1013 hPa)
Melting Pt: 148…152 °C
Density: 1.426 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00008325
CAS Number: 137-26-8
EINECS: 205-286-2
UN: 2811
ADR: 6.1,II
Merck Index: 12,09510

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR