1,2,4,5-Tetramethylbenzene ≥98%

T19607-100G T19607-500G
220226-121EA 70.22 USD
220226-121 220226-122
1,2,4,5-Tetramethylbenzene ≥98%
1,2,4,5-Tetramethylbenzene
Formula: C₁₀H₁₄
MW: 134.22 g/mol
Boiling Pt: 196.8 °C (1013 hPa)
Melting Pt: 78 - 81 °C
Density: 1.036 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008528
CAS Number: 95-93-2
EINECS: 202-465-7
UN: 1325
ADR: 4.1,II
Merck Index: 13,03501

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR