2,2',4,4'-Tetrahydroxybenzophenone ≥97%

Synonyms: Bis(2,4-dihydroxyphenyl)methanone

T16403-25G T16403-100G
220226-024EA 64.62 USD
220226-024 220226-023
2,2',4,4'-Tetrahydroxybenzophenone ≥97%
2,2',4,4'-Tetrahydroxybenzophenone
Formula: C₁₃H₁₀O₅
MW: 246.22 g/mol
Boiling Pt: 100…102 °C (40 mmHg)
Melting Pt: 196…198 °C
Density: 1.498 g/cm³ (20 °C)
MDL Number: MFCD00002278
CAS Number: 131-55-5
EINECS: 205-028-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR