1,2,3,4-Tetrahydroquinoline

T15504-500G T15504-5G T15504-100G
220225-520EA 197.08 USD
220225-520 220226-003 220225-519
1,2,3,4-Tetrahydroquinoline
1,2,3,4-Tetrahydroquinoline
Formula: C₉H₁₁N
MW: 133.19 g/mol
MDL Number: MFCD00006693
CAS Number: 635-46-1
EINECS: 211-237-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR