Thiophene-2-carboxylic hydrazide

Synonyms: 2-Thiophenecarboxylic hydrazide, 2-Thiophenecarboxylic acid hydrazide, Thiophene-2-carbohydrazide

T1388-1G T1388-5G T1388-25G
220225-791EA 45.23 USD
220225-791 220225-793 220225-792
Thiophene-2-carboxylic hydrazide
Thiophene-2-carboxylic hydrazide
Formula: C₅H₆N₂OS
MW: 142.18 g/mol
Melting Pt: 142…143 °C
MDL Number: MFCD00005435
CAS Number: 2361-27-5
EINECS: 219-107-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR