2,4,6-Tris(2-pyridyl)-1,3,5-triazine

Synonyms: 2,4,6-Tri(2-pyridyl)-s-triazine, 2,4,6-Tri(2-pyridyl)-1,3,5-triazine, 2,4,6-Tris(2-pyridyl)-s-triazine

T1253-10G T1253-25G T1253-5G T1253-1G
220225-960EA 572.31 USD
220225-960 220225-962 220225-963 220225-961
2,4,6-Tris(2-pyridyl)-1,3,5-triazine
2,4,6-Tris(2-pyridyl)-1,3,5-triazine
Formula: C₁₈H₁₂N₆
MW: 312.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006045
CAS Number: 3682-35-7
EINECS: 222-965-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cae28c9fe6b63cdfc9b8d64479524c4e0d2ca9d&vid=8513f4cb36c0a272d4a4e3ea99f42e1c07aedf92&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=562efd6672c9772350cba16c6b48f9c529dfa426&vid=c9c956055af0b012dfc5454df2609376fc0ecd74&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fae539280a938bda6014c19f2fc631c41828b2cd&vid=b60f1b6c3e35c4c9cde8c90b7757c8720d093eee&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=780a1b89197c24bf6f32fe387d85ca33947d5ad8&vid=82e8d71a025aa0076178e7107537e18f62d38fff&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR