Sodium acetate trihydrate ≥99.0%

S7670-250G S7670-5KG S7670-500G S7670-1KG
220210-237EA 52.43 USD
220210-237 220210-239 220210-238 220210-236
Sodium acetate trihydrate ≥99.0%
Sodium acetate trihydrate
Formula: H₃CCOONa·3H₂O
Boiling Pt: 120…123 °C (1013 hPa)
Melting Pt: 58 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=595fbd8527de098592b429140a59fb0d415a73ed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR