Syringic acid

Synonyms: 4-Hydroxy-3,5-dimethoxybenzoic acid

S6881-100G S6881-10G S6881-25G S6881-5G
220209-232EA 1078.85 USD
220209-232 220209-233 220209-234 220209-235
Syringic acid
Syringic acid
Formula: C₉H₁₀O₅
MW: 198.18 g/mol
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 204…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00002552
CAS Number: 530-57-4
EINECS: 208-486-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR