Methyl stearate ≥99% (by GC)

Synonyms: Stearic acid methyl ester, Methyl octadecanoate

S5376-10G S5376-50G S5376-1G S5376-5G
220209-039EA 251.08 USD
220209-039 220208-066 220208-065 220208-067
Methyl stearate ≥99% (by GC)
Methyl stearate
Formula: CH₃(CH₂)₁₆CO₂CH₃
MW: 298.51 g/mol
Boiling Pt: 355 °C (1013 hPa)
Melting Pt: 37…39 °C
Density: 1.007 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009005
CAS Number: 112-61-8
EINECS: 203-990-4
Merck Index: 12,08959

Order Now

Specification Test Results

Assay ~99 % (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR