Suberic acid ≥98%

Warning

Synonyms: 1,8-Octanedioic acid, Octanedioic acid, 1,6-Hexanedicarboxylic acid

S5200-5G S5200-500G S5200-100G
220208-803EA 67.74 USD
220208-803 220208-802 220209-620
Suberic acid ≥98%
Suberic acid
Formula: C₈H₁₄O₄
MW: 174.2 g/mol
Boiling Pt: 361 °C (1013 hPa)
Melting Pt: 139…142 °C
Density: 1.354 g/cm³ (21 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004428
CAS Number: 505-48-6
EINECS: 208-010-9
Merck Index: 12,09031

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR