cis-Stilbene ≥96%

Synonyms: cis-1,2-Diphenylethene, Isostilbene, cis-1,2-Diphenylethylene, (Z)-1,2-Diphenylethene

S4808-10G S4808-1G S4808-5G
220209-457EA 276.77 USD
220209-457 220209-458 220209-459
cis-Stilbene ≥96%
cis-Stilbene
Formula: (C₆H₅)CHCH(C₆H₅)
MW: 180.25 g/mol
Boiling Pt: 307 °C (1013 hPa)
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004788
CAS Number: 645-49-8
EINECS: 211-445-7
Merck Index: 12,08972

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR